4-Bromo-2-fluorophenylboronic acid N-butyldiethanolamine ester ≥97%

Supplier: AOBChem USA

Warning

19437-5G 19437-1G 19437-25G 19437-500MG
77634-276EA 295 USD
77634-276 77637-104 77657-868 77645-106
4-Bromo-2-fluorophenylboronic acid N-butyldiethanolamine ester ≥97%
4-Bromo-2-fluorophenylboronic acid N-butyldiethanolamine ester

Precaución: For Research Use Only

Formula: 344.028 g/mol
MW: C₁₄H₂₀BBrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD19105363
CAS Number: 1451391-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR