2-Chloro-5-isopropylbenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
19416-5G
19416-1G
19416-25G
19416-500MG
77631-924EA
1430
USD
77631-924
77637-096
77657-858
77645-102
2-Chloro-5-isopropylbenzaldehyde ≥97%
2-Chloro-5-isopropylbenzaldehyde
Precaución: For Research Use Only
Formula:
182.647 g/mol MW: C₁₀H₁₁ClO Storage Temperature: Ambient |
MDL Number:
MFCD29119800 CAS Number: 1288991-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...