2-Chloro-4-fluoro-3-methyl-1-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

194077-5G 194077-1G 194077-25G 194077-500MG
77631-922EA 1288 USD
77631-922 77637-094 77657-856 77645-100
2-Chloro-4-fluoro-3-methyl-1-(trifluoromethyl)benzene ≥95%
2-Chloro-4-fluoro-3-methyl-1-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: 212.57 g/mol
MW: C₈H₅ClF₄
Storage Temperature: Ambient
MDL Number: MFCD28751913
CAS Number: 1056942-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR