4-Iodo-2,6-dimethylpyridine ≥97%

Supplier: AOBChem USA

Warning

19374-1G 19374-5G 19374-500MG
77637-092EA 531 USD
77637-092 77654-040 77649-138
4-Iodo-2,6-dimethylpyridine ≥97%
4-Iodo-2,6-dimethylpyridine

Precaución: For Research Use Only

Formula: C₇H₈IN
Storage Temperature: Ambient
MDL Number: MFCD12924838
CAS Number: 22282-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR