2,4-Dibromo-5-methylphenol ≥97%

Supplier: AOBChem USA

Warning

193711-5G 193711-1G 193711-25G 193711-500MG
77631-920EA 1055 USD
77631-920 77637-090 77657-854 77645-098
2,4-Dibromo-5-methylphenol ≥97%
2,4-Dibromo-5-methylphenol

Precaución: For Research Use Only

Formula: 265.93 g/mol
MW: C₇H₆Br₂O
Storage Temperature: Ambient
MDL Number: MFCD33484630
CAS Number: 13321-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR