2,4,5-Tribromophenol ≥97%

Supplier: AOBChem USA

Warning

193708-5G 193708-1G 193708-500MG
77628-028EA 1484.23 USD
77628-028 77641-090 77645-096
2,4,5-Tribromophenol ≥97%
2,4,5-Tribromophenol

Precaución: For Research Use Only

Formula: C₆H₃Br₃O
Storage Temperature: Ambient
MDL Number: MFCD09037674
CAS Number: 14401-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR