1-Bromo-4-methoxy-2-methylsulfanylbenzene ≥95%

Supplier: AOBChem USA

Warning

19358-1G 19358-500MG
77634-272EA 2242 USD
77634-272 77649-136
1-Bromo-4-methoxy-2-methylsulfanylbenzene ≥95%
1-Bromo-4-methoxy-2-methylsulfanylbenzene

Precaución: For Research Use Only

Formula: 233.125 g/mol
MW: C₈H₉BrOS
Storage Temperature: Ambient
MDL Number: MFCD11043740
CAS Number: 72856-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR