2-Bromo-3-iodothiophene ≥97%

Supplier: AOBChem USA

Warning

19310-1G 19310-500MG 19310-5G
77637-086EA 747 USD
77637-086 77652-438 77657-852
2-Bromo-3-iodothiophene ≥97%
2-Bromo-3-iodothiophene

Precaución: For Research Use Only

Formula: 288.932 g/mol
MW: C₄H₂BrIS
Storage Temperature: Ambient
MDL Number: MFCD08234871
CAS Number: 24287-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR