3-Acetyl-2-bromothiophene ≥95%

Supplier: AOBChem USA

Warning

19308-5G 19308-25G 19308-1G 19308-500MG
77631-918EA 816 USD
77631-918 77654-036 77645-092 77645-094
3-Acetyl-2-bromothiophene ≥95%
3-Acetyl-2-bromothiophene

Precaución: For Research Use Only

Formula: C₆H₅BrOS
Storage Temperature: Ambient
MDL Number: MFCD25958595
CAS Number: 137272-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR