Tert-butyl (2,6-dichloro-4-(trifluoromethyl)phenyl)carbamate ≥95%

Supplier: AOBChem USA

Warning

193041-5G 193041-25G 193041-1G 193041-500MG
77628-024EA 1703.62 USD
77628-024 77635-516 77637-084 77645-090
Tert-butyl (2,6-dichloro-4-(trifluoromethyl)phenyl)carbamate ≥95%
Tert-butyl (2,6-dichloro-4-(trifluoromethyl)phenyl)carbamate

Precaución: For Research Use Only

Formula: C₁₂H₁₂Cl₂F₃NO₂
Storage Temperature: Ambient
CAS Number: 3031792-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR