5-Cyanothiophene-2-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

19286-5G 19286-25G 19286-1G 19286-500MG
77631-916EA 839 USD
77631-916 77654-032 77641-088 77645-088
5-Cyanothiophene-2-carboxylic acid ≥95%
5-Cyanothiophene-2-carboxylic acid

Precaución: For Research Use Only

Formula: C₆H₃NO₂S
Storage Temperature: Ambient
MDL Number: MFCD12027190
CAS Number: 59786-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR