2-Bromo-5-isopropylpyridine ≥95%

Supplier: AOBChem USA

Warning

19273-5G 19273-25G 19273-1G 19273-500MG
77634-270EA 380 USD
77634-270 77654-030 77641-086 77649-134
2-Bromo-5-isopropylpyridine ≥95%
2-Bromo-5-isopropylpyridine

Precaución: For Research Use Only

Formula: C₈H₁₀BrN
Storage Temperature: Ambient
MDL Number: MFCD11869553
CAS Number: 1142197-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR