1-Bromo-2-fluoro-3-propoxybenzene ≥95%

Supplier: AOBChem USA

Warning

192685-5G 192685-1G 192685-25G 192685-500MG
77631-914EA 760 USD
77631-914 77637-082 77654-028 77649-132
1-Bromo-2-fluoro-3-propoxybenzene ≥95%
1-Bromo-2-fluoro-3-propoxybenzene

Precaución: For Research Use Only

Formula: C₉H₁₀BrFO
MW: 233.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29043783
CAS Number: 1881329-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR