2,2-Dibromo-1-(4-iodophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

192275-1G 192275-5G 192275-500MG
77634-266EA 1078 USD
77634-266 77654-022 77649-126
2,2-Dibromo-1-(4-iodophenyl)ethanone ≥95%
2,2-Dibromo-1-(4-iodophenyl)ethanone

Precaución: For Research Use Only

Formula: 403.84 g/mol
MW: C₈H₅Br₂IO
Storage Temperature: Ambient
MDL Number: MFCD09743654
CAS Number: 556110-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR