2-Bromo-4'-(methylthio)acetophenone ≥95%

Supplier: AOBChem USA

Warning

192235-5G 192235-25G
77634-264EA 123 USD
77634-264 77657-848
2-Bromo-4'-(methylthio)acetophenone ≥95%
2-Bromo-4'-(methylthio)acetophenone

Precaución: For Research Use Only

Formula: C₉H₉BrOS
Storage Temperature: Ambient
MDL Number: MFCD03094708
CAS Number: 42445-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR