2-(3,5-Dimethylphenoxy)acetonitrile ≥97%

Supplier: AOBChem USA

Warning

191842-5G 191842-1G 191842-500MG
77657-844EA 2317 USD
77657-844 77641-082 77645-086
2-(3,5-Dimethylphenoxy)acetonitrile ≥97%
2-(3,5-Dimethylphenoxy)acetonitrile

Precaución: For Research Use Only

Formula: 161.2 g/mol
MW: C₁₀H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD09906402
CAS Number: 33901-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR