4-(2,4-Dichlorophenoxy)benzonitrile ≥95%

Supplier: AOBChem USA

191801-5G 191801-1G 191801-500MG
77657-842EA 1828 USD
77657-842 77641-080 77645-084
4-(2,4-Dichlorophenoxy)benzonitrile ≥95%
4-(2,4-Dichlorophenoxy)benzonitrile

Precaución: For Research Use Only

Formula: C₁₃H₇Cl₂NO
MDL Number: MFCD01815091
CAS Number: 24789-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR