4-(2,4-Dichlorophenoxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

191792-5G 191792-25G 191792-1G 191792-500MG
77634-262EA 323 USD
77634-262 77657-840 77641-078 77649-124
4-(2,4-Dichlorophenoxy)benzaldehyde ≥95%
4-(2,4-Dichlorophenoxy)benzaldehyde

Precaución: For Research Use Only

Formula: C₁₃H₈Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD08276810
CAS Number: 78725-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR