1-(2,3-Difluoro-4-methylphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

191687-5G 191687-1G 191687-500MG
77628-020EA 1484.23 USD
77628-020 77637-078 77645-082
1-(2,3-Difluoro-4-methylphenyl)-1-propanone ≥95%
1-(2,3-Difluoro-4-methylphenyl)-1-propanone

Precaución: For Research Use Only

Formula: 184.18 g/mol
MW: C₁₀H₁₀F₂O
Storage Temperature: Ambient
MDL Number: MFCD30087326
CAS Number: 1694387-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR