N,N-Diisopropyl-3-methylbenzamide ≥95%

Supplier: AOBChem USA

Warning

191109-5G 191109-1G 191109-500MG
77654-016EA 2622 USD
77654-016 77641-072 77645-074
N,N-Diisopropyl-3-methylbenzamide ≥95%
N,N-Diisopropyl-3-methylbenzamide

Precaución: For Research Use Only

Formula: C₁₄H₂₁NO
Storage Temperature: Ambient
MDL Number: MFCD00587389
CAS Number: 5448-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR