3-Bromo-2,4,6-trimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

19078-5G 19078-25G 19078-1G 19078-500MG
77631-910EA 1350 USD
77631-910 77657-836 77645-072 77649-118
3-Bromo-2,4,6-trimethylbenzaldehyde ≥97%
3-Bromo-2,4,6-trimethylbenzaldehyde

Precaución: For Research Use Only

Formula: C₁₀H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD30572266
CAS Number: 88174-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR