3-Chloro-4-fluoro-α-(fluoromethyl)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

190205-5G 190205-1G 190205-500MG
77628-016EA 1553.24 USD
77628-016 77641-068 77645-068
3-Chloro-4-fluoro-α-(fluoromethyl)benzenemethanol ≥97%
3-Chloro-4-fluoro-α-(fluoromethyl)benzenemethanol

Precaución: For Research Use Only

Formula: 192.59 g/mol
MW: C₈H₇ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD34861050
CAS Number: 2228407-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR