3-Bromo-2-(pyrrolidin-1-yl)pyridine ≥95%
Supplier: AOBChem USA
|
Warning
|
18989-5G
18989-25G
18989-1G
77634-258EA
224
USD
77634-258
77654-010
77641-066
3-Bromo-2-(pyrrolidin-1-yl)pyridine ≥95%
3-Bromo-2-(pyrrolidin-1-yl)pyridine
Precaución: For Research Use Only
Formula:
227.101 g/mol MW: C₉H₁₁BrN₂ Storage Temperature: Ambient |
MDL Number:
MFCD12026111 CAS Number: 1150561-84-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...