1,1-Dimethylethyl 3-(5-fluoro-2-methylphenyl)-3-hydroxy-1-azetidinecarboxylate ≥95%
Supplier: AOBChem USA
|
Warning
|
189259-5G
189259-1G
189259-500MG
77628-010EA
1484.23
USD
77628-010
77637-072
77645-060
1,1-Dimethylethyl 3-(5-fluoro-2-methylphenyl)-3-hydroxy-1-azetidinecarboxylate ≥95%
1,1-Dimethylethyl 3-(5-fluoro-2-methylphenyl)-3-hydroxy-1-azetidinecarboxylate
Precaución: For Research Use Only
Formula:
281.32 g/mol MW: C₁₅H₂₀FNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD34860650 CAS Number: 1484142-37-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...