1,1-Dimethylethyl 3-(2-fluoro-5-methylphenyl)-3-hydroxy-1-azetidinecarboxylate ≥95%

Supplier: AOBChem USA

Warning

189252-5G 189252-25G 189252-1G 189252-500MG
77631-904EA 1368 USD
77631-904 77654-008 77641-058 77645-058
1,1-Dimethylethyl 3-(2-fluoro-5-methylphenyl)-3-hydroxy-1-azetidinecarboxylate ≥95%
1,1-Dimethylethyl 3-(2-fluoro-5-methylphenyl)-3-hydroxy-1-azetidinecarboxylate

Precaución: For Research Use Only

Formula: 281.32 g/mol
MW: C₁₅H₂₀FNO₃
Storage Temperature: Ambient
MDL Number: MFCD30802684
CAS Number: 1493550-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR