1,1-Dimethylethyl 3-hydroxy-3-(4-methylphenyl)-1-azetidinecarboxylate ≥95%

Supplier: AOBChem USA

Warning

189170-5G 189170-25G 189170-1G 189170-500MG
77631-902EA 1410 USD
77631-902 77657-828 77641-056 77649-114
1,1-Dimethylethyl 3-hydroxy-3-(4-methylphenyl)-1-azetidinecarboxylate ≥95%
1,1-Dimethylethyl 3-hydroxy-3-(4-methylphenyl)-1-azetidinecarboxylate

Precaución: For Research Use Only

Formula: 263.33 g/mol
MW: C₁₅H₂₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD20864846
CAS Number: 1472706-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR