1,1-Dimethylethyl 3-hydroxy-3-(3-methylphenyl)-1-azetidinecarboxylate ≥95%

Supplier: AOBChem USA

Warning

189168-5G 189168-1G 189168-25G 189168-500MG
77631-900EA 1175 USD
77631-900 77637-068 77654-006 77645-054
1,1-Dimethylethyl 3-hydroxy-3-(3-methylphenyl)-1-azetidinecarboxylate ≥95%
1,1-Dimethylethyl 3-hydroxy-3-(3-methylphenyl)-1-azetidinecarboxylate

Precaución: For Research Use Only

Formula: 263.33 g/mol
MW: C₁₅H₂₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD21596778
CAS Number: 1490999-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR