Tert-butyl 3-hydroxy-3-[2-(trifluoromethoxy)phenyl]azetidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

189164-5G 189164-25G 189164-1G 189164-500MG
77631-898EA 1427 USD
77631-898 77654-004 77641-052 77645-050
Tert-butyl 3-hydroxy-3-[2-(trifluoromethoxy)phenyl]azetidine-1-carboxylate ≥95%
Tert-butyl 3-hydroxy-3-[2-(trifluoromethoxy)phenyl]azetidine-1-carboxylate

Precaución: For Research Use Only

Formula: C₁₅H₁₈F₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD30802716
CAS Number: 1609671-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR