1,1-Dimethylethyl 3-(3-fluorophenyl)-3-hydroxy-1-azetidinecarboxylate ≥95%

Supplier: AOBChem USA

Warning

189134-5G 189134-1G 189134-25G 189134-500MG
77631-896EA 988 USD
77631-896 77637-066 77657-826 77649-112
1,1-Dimethylethyl 3-(3-fluorophenyl)-3-hydroxy-1-azetidinecarboxylate ≥95%
1,1-Dimethylethyl 3-(3-fluorophenyl)-3-hydroxy-1-azetidinecarboxylate

Precaución: For Research Use Only

Formula: 267.3 g/mol
MW: C₁₄H₁₈FNO₃
Storage Temperature: Ambient
MDL Number: MFCD21081303
CAS Number: 1380752-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR