1-Benzyl-3-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

189089-5G 189089-25G 189089-1G 189089-500MG
77628-006EA 1797.35 USD
77628-006 77657-824 77641-050 77645-046
1-Benzyl-3-methoxybenzene ≥95%
1-Benzyl-3-methoxybenzene

Precaución: For Research Use Only

Formula: 198.2603 g/mol
MW: C₁₄H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD34860552
CAS Number: 23450-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR