3-Bromo-2,4-dichlorophenol ≥97%

Supplier: AOBChem USA

Warning

189080-5G 189080-25G 189080-1G 189080-500MG
77631-892EA 1400 USD
77631-892 77657-822 77641-048 77649-108
3-Bromo-2,4-dichlorophenol ≥97%
3-Bromo-2,4-dichlorophenol

Precaución: For Research Use Only

Formula: 241.8974 g/mol
MW: C₆H₃BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD28737849
CAS Number: 13659-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR