2-Methoxy-6-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

18901-5G 18901-25G
77634-256EA 158 USD
77634-256 77657-820
2-Methoxy-6-(trifluoromethyl)benzoic acid ≥97%
2-Methoxy-6-(trifluoromethyl)benzoic acid

Precaución: For Research Use Only

Formula: C₉H₇F₃O₃
MW: 220.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832319
CAS Number: 119692-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR