Tert-butyl 3-ethoxy-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

188779-5G 188779-25G 188779-1G 188779-500MG
77628-004EA 1524.4 USD
77628-004 77635-514 77637-064 77645-042
Tert-butyl 3-ethoxy-2-methylbenzoate ≥97%
Tert-butyl 3-ethoxy-2-methylbenzoate

Precaución: For Research Use Only

Formula: 236.307 g/mol
MW: C₁₄H₂₀O₃
Storage Temperature: Ambient
MDL Number: MFCD34860369
CAS Number: 2969346-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR