2-Methoxy-5-(trifluoromethyl)-3-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

18847-5G 18847-1G 18847-25G 18847-500MG
77634-254EA 265 USD
77634-254 77637-062 77657-818 77649-106
2-Methoxy-5-(trifluoromethyl)-3-pyridineboronic acid ≥97%
2-Methoxy-5-(trifluoromethyl)-3-pyridineboronic acid

Precaución: For Research Use Only

Formula: C₇H₇BF₃NO₃
MW: 220.942 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10696251
CAS Number: 1072946-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR