1-Bromo-3-chloro-4-methoxy-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

18751-5G 18751-25G 18751-1G 18751-500MG
77631-886EA 1117 USD
77631-886 77657-814 77641-042 77649-098
1-Bromo-3-chloro-4-methoxy-5-(trifluoromethyl)benzene ≥97%
1-Bromo-3-chloro-4-methoxy-5-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: 289.477 g/mol
MW: C₈H₅BrClF₃O
Storage Temperature: Ambient
MDL Number: MFCD28015430
CAS Number: 1809161-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR