4-(3-Bromophenyl)-1-(phenylmethyl)-4-piperidinol ≥95%

Supplier: AOBChem USA

Warning

187096-5G 187096-1G 187096-25G 187096-500MG
77631-884EA 1206 USD
77631-884 77637-050 77657-812 77645-038
4-(3-Bromophenyl)-1-(phenylmethyl)-4-piperidinol ≥95%
4-(3-Bromophenyl)-1-(phenylmethyl)-4-piperidinol

Precaución: For Research Use Only

Formula: 346.26 g/mol
MW: C₁₈H₂₀BrNO
Storage Temperature: Ambient
MDL Number: MFCD34859289
CAS Number: 157133-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR