3,5-Dimethyl-2-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-3,5-dimethylpyridine

18707-5G 18707-1G 18707-25G 18707-500MG
77634-246EA 349 USD
77634-246 77637-048 77657-810 77645-036
3,5-Dimethyl-2-fluoropyridine ≥97%
3,5-Dimethyl-2-fluoropyridine

Precaución: For Research Use Only

Formula: C₇H₈FN
MW: 125.144 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044345
CAS Number: 111887-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR