α-Cyclohexyl-4-propylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186979-5G 186979-25G 186979-1G 186979-500MG
77627-998EA 1777.78 USD
77627-998 77635-512 77641-040 77645-034
α-Cyclohexyl-4-propylbenzenemethanol ≥95%
α-Cyclohexyl-4-propylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₆H₂₄O
Storage Temperature: Ambient
MDL Number: MFCD14530734
CAS Number: 1216001-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR