α-Cyclohexyl-2,4-difluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186964-5G 186964-1G 186964-500MG
77627-996EA 1553.24 USD
77627-996 77641-038 77645-032
α-Cyclohexyl-2,4-difluorobenzenemethanol ≥95%
α-Cyclohexyl-2,4-difluorobenzenemethanol

Precaución: For Research Use Only

Formula: C₁₃H₁₆F₂O
Storage Temperature: Ambient
MDL Number: MFCD12787303
CAS Number: 1249419-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR