α-Cyclohexyl-3-(trifluoromethoxy)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186963-5G 186963-25G 186963-1G 186963-500MG
77627-994EA 1777.78 USD
77627-994 77635-510 77637-044 77645-030
α-Cyclohexyl-3-(trifluoromethoxy)benzenemethanol ≥95%
α-Cyclohexyl-3-(trifluoromethoxy)benzenemethanol

Precaución: For Research Use Only

Formula: C₁₄H₁₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD17406642
CAS Number: 1272965-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR