Benzenemethanol, α-cyclohexyl-2-ethyl- ≥95%

Supplier: AOBChem USA

Warning

186932-5G 186932-25G 186932-1G 186932-500MG
77627-990EA 1583.11 USD
77627-990 77635-506 77637-040 77649-088
Benzenemethanol, α-cyclohexyl-2-ethyl- ≥95%
Benzenemethanol, α-cyclohexyl-2-ethyl-

Precaución: For Research Use Only

Formula: 218.34 g/mol
MW: C₁₅H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD19602031
CAS Number: 1486394-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR