2-Chloro-α-cyclohexyl-4-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186895-5G 186895-25G 186895-1G 186895-500MG
77627-988EA 1583.11 USD
77627-988 77657-806 77641-036 77645-028
2-Chloro-α-cyclohexyl-4-methylbenzenemethanol ≥95%
2-Chloro-α-cyclohexyl-4-methylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₄H₁₉ClO
Storage Temperature: Ambient
MDL Number: MFCD35167097
CAS Number: 1702891-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR