Benzenemethanol, α-cyclohexyl-2-(1-methylethyl)- ≥97%

Supplier: AOBChem USA

Warning

186858-5G 186858-25G 186858-1G 186858-500MG
77627-986EA 1484.23 USD
77627-986 77635-504 77637-038 77649-086
Benzenemethanol, α-cyclohexyl-2-(1-methylethyl)- ≥97%
Benzenemethanol, α-cyclohexyl-2-(1-methylethyl)-

Precaución: For Research Use Only

Formula: C₁₆H₂₄O
Storage Temperature: Ambient
MDL Number: MFCD34859239
CAS Number: 2273817-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR