α-Cyclohexyl-3,4-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186850-5G 186850-25G 186850-1G 186850-500MG
77627-984EA 1484.23 USD
77627-984 77635-502 77641-034 77649-084
α-Cyclohexyl-3,4-dimethylbenzenemethanol ≥95%
α-Cyclohexyl-3,4-dimethylbenzenemethanol

Precaución: For Research Use Only

Formula: 218.34 g/mol
MW: C₁₅H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD11937137
CAS Number: 872289-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR