α-Cyclohexyl-3-(trifluoromethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186848-5G 186848-1G 186848-500MG
77657-804EA 2250 USD
77657-804 77641-032 77645-026
α-Cyclohexyl-3-(trifluoromethyl)benzenemethanol ≥95%
α-Cyclohexyl-3-(trifluoromethyl)benzenemethanol

Precaución: For Research Use Only

Formula: C₁₄H₁₇F₃O
Storage Temperature: Ambient
MDL Number: MFCD12605270
CAS Number: 1183567-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR