α-Cyclohexyl-3-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186847-5G 186847-25G 186847-1G 186847-500MG
77627-982EA 1797.35 USD
77627-982 77635-500 77637-036 77649-082
α-Cyclohexyl-3-methoxybenzenemethanol ≥95%
α-Cyclohexyl-3-methoxybenzenemethanol

Precaución: For Research Use Only

Formula: C₁₄H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD12153458
CAS Number: 1207204-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR