α-Cyclohexyl-4-(trifluoromethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186844-5G 186844-1G 186844-500MG
77627-980EA 1484.23 USD
77627-980 77637-034 77649-080
α-Cyclohexyl-4-(trifluoromethyl)benzenemethanol ≥95%
α-Cyclohexyl-4-(trifluoromethyl)benzenemethanol

Precaución: For Research Use Only

Formula: C₁₄H₁₇F₃O
Storage Temperature: Ambient
MDL Number: MFCD16293521
CAS Number: 888722-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR