α-Cyclohexyl-4-(dimethylamino)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186838-5G 186838-25G 186838-1G 186838-500MG
77627-978EA 1553.24 USD
77627-978 77635-498 77641-030 77645-024
α-Cyclohexyl-4-(dimethylamino)benzenemethanol ≥95%
α-Cyclohexyl-4-(dimethylamino)benzenemethanol

Precaución: For Research Use Only

Formula: 233.35 g/mol
MW: C₁₅H₂₃NO
Storage Temperature: Ambient
MDL Number: MFCD16317269
CAS Number: 101355-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR