α-Cyclohexyl-2-naphthalenemethanol ≥95%

Supplier: AOBChem USA

Warning

186836-5G 186836-25G 186836-1G 186836-500MG
77627-976EA 1484.23 USD
77627-976 77635-496 77645-022 77649-078
α-Cyclohexyl-2-naphthalenemethanol ≥95%
α-Cyclohexyl-2-naphthalenemethanol

Precaución: For Research Use Only

Formula: C₁₇H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD06654251
CAS Number: 186407-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR