α-Cyclohexyl-4-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186832-5G 186832-25G 186832-1G 186832-500MG
77631-882EA 938 USD
77631-882 77653-988 77641-028 77649-076
α-Cyclohexyl-4-methylbenzenemethanol ≥95%
α-Cyclohexyl-4-methylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₄H₂₀O
Storage Temperature: Ambient
MDL Number: MFCD11935946
CAS Number: 28047-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR